Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medical Action™ Low-Density Institutional Trash Can Liners Available on GSA/VA Contract for Federal Government customers only.

Variety of sizes, thicknesses and colors for waste disposal needs

Manufacturer:  American Precision Plastics R4651

 View more versions of this product

Catalog No. 14-828-216

  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

LLDPE (Linear Low-density Polyethylene)
96.5 x 114.3cm (38 x 45 in.)
For waste disposal
96.5cm
0.95 mil
44 gal.
Brown
114.3cm
Yes
38 in.
Trash Bag, Coreless Roll
45 in.
166L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit