Medical Action™ Low-Density Laundry and Linen Bags in Rolls

Manufacturer: American Precision Plastics R112M

 View more versions of this product

Catalog No. 14-828-196

  Check Availability
Add to Cart

    Specifications

    Product Type Laundry and Linen Bag
    Capacity (English) 20 to 30 gal.
    Color Yellow, Red
    Guage (English) 1.1mil
    For Use With (Application) Laundry applications
    Includes Infectious linen, biohazard with biohazard symbol
    Capacity 20 to 30 gal., 75 to 113L
    Dimensions (L x W) 30.5 x 41 in. (77.4 x 104.1cm)
    Material LLDPE
    Length (Metric) 77.4cm
    Length (English) 30.5 in.
    Width (Metric) 104.1cm
    Width (English) 41 in.
    Capacity (Metric) 75 to 113L

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit