Learn More
Medical Packaging Pap-Pak Kit with the Papette Cervical Cell Collector
The Wallach Papette collects all cells essential for tracing the early stages of cervical cancer
$126.25
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
22-037749
![]() |
Medical Packaging
CSW 1 |
Full-spectrum Cervical Broom Cell Collector |
Pack of 100 for $126.25
Case of 10 PK for $1,262.46 |
|
Description
- By inserting into the cervical canal and rotating once or twice while applying light pressure, the 8 in. (20cm) sampler collects endocervical, ectocervical, and T-zone cells
- The soft, flexible polyethylene bristles of the Papette provide enhanced pressure control and prevent internal injury, which can result in smears obscured by blood during sampling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.