Medical Packaging Pap-Pak™ Pap Smear Collection System

Each kit contains all the necessary materials for specimen collection, smear preparation, fixation and transportation to the laboratory

Manufacturer: Medical PackagingSupplier Diversity Partner S100

 View more versions of this product

Catalog No. 22-383380

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) For endocervical specimen collection
    Includes Cell Scraper, Cotton Swab, Foil Pouch Fixative and Single Frosted Slide
    Quantity 25 Pack
    Sample Type Vaginal, cervical and endocervical smears
    Product Type Pap Smear Endocervical Specimen Collection Kit without mailing bracket

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit