Learn More
Medical Packaging Pap-Pak™ Pap Smear Collection System Available on GSA/VA Contract for Federal Government customers only.
Each kit contains all the necessary materials for specimen collection, smear preparation, fixation and transportation to the laboratory
Manufacturer:  Medical PackagingSupplier Diversity Partner C100P1
 View more versions of this product
Catalog No. 22-037738
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
For endocervical specimen collection
25 Pack
Pap Smear Endocervical Specimen Collection Kit without mailing bracket
Cytology Brush Pap-Pak Kits with 8 in. CytoSoft Cytology Brush, Cell Scraper, Pump Spray Fixative and Single Frosted Slide
Vaginal, cervical and endocervical smears
Accessories