Medical Wire Transwab™ Amies Transport System for Aerobes and Anaerobes: Amies Duo

Self-contained transport swab with semisolid medium

Manufacturer: ADVANTAGE BUNDLING SP LLC MW169C

 View more versions of this product

Catalog No. 23-691-022

  Check Availability
Add to Cart

    Specifications

    Product Type Transwab™ Amies Charcoal
    Closure Color White
    Shelf Life 2 Year(s)
    Certifications/Compliance CE, MDD, IVD, CLSI M40
    For Use With (Application) Wound, skin, urogenital, throat, and multiple sites
    Length (English) 5.9 in.
    Length (Metric) 15cm
    Quantity 125 Pack
    Shaft Plastic
    Swab Configuration Double

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit