Medical Wire Transwab™ Amies Transport System for Aerobes and Anaerobes: Amies

Self-contained transport swab with semisolid medium

Manufacturer: ADVANTAGE BUNDLING SP LLC MW170

 View more versions of this product

Catalog No. 23-691-001

  Check Availability
Add to Cart

    Specifications

    Product Type Transwab™ Amies
    Closure Color Blue
    Shelf Life 2 Year(s)
    Certifications/Compliance CE, MDD, IVD, CLSI M40
    For Use With (Application) Wound, skin, urogenital and throat
    Length (English) 5.9 in.
    Length (Metric) 15cm
    Quantity 125 Pack
    Shaft Plastic
    Swab Configuration Single

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit