Learn More
Medical Wire Transwab™ Amies Transport System for Aerobes and Anaerobes: Pernasal Amies
Self-contained transport swab with semisolid medium
Manufacturer:  ADVANTAGE BUNDLING SP LLC MW173C
 View more versions of this product
Catalog No. 23-691-006
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light Blue
125 Pack
Single
Nasopharyngeal, pediatric
15cm
Transwab™ Pernasal Amies Charcoal
Ultrafine twisted wire
CE, MDD, IVD, CLSI M40
5.9 in.
Safety and Handling
ShelfLife : 2 Year(s)