Medical Wire Transwab™ Amies Transport System for Aerobes and Anaerobes: Pernasal Amies

Self-contained transport swab with semisolid medium

Manufacturer: ADVANTAGE BUNDLING SP LLC MW173P

 View more versions of this product

Catalog No. 23-691-005

  Check Availability
Add to Cart

    Specifications

    Product Type Transwab™ Pernasal Amies
    Closure Color Light Blue
    Shelf Life 2 Year(s)
    Certifications/Compliance CE, MDD, IVD, CLSI M40
    For Use With (Application) Nasopharyngeal, pediatric
    Length (English) 5.9 in.
    Length (Metric) 15cm
    Quantity 125 Pack
    Shaft Ultrafine twisted wire
    Swab Configuration Single

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit