MediPurpose™ Safety Lancet

Safety needle or blade lancet is safely concealed before and after use

Manufacturer: MEDIPURPOSE SLB200

 View more versions of this product

Catalog No. 22-000-103

  Check Availability
Add to Cart

    Specifications

    Capacity (Metric) 75 to 100μL
    Capacity 0.0025 to 0.00338 oz., 75 to 100μL
    Color Green
    Depth (English) 0.7 in.
    Depth (Metric) 1.8mm
    For Use With (Application) For all POC tests requiring whole Blood samples, including Glucose, cholesterol, HIV and more
    Includes Blade
    Needle Gauge 19g
    Quantity 100 Pack
    Product Type Safety Lancet
    Capacity (English) 0.0025 to 0.00338 oz.
    Certifications/Compliance OSHA

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit