Medique™ Medi-First™ Non Aspirin Extra-Strength

Manufacturer: Moore Medical Corp 64101

 View more versions of this product

Catalog No. 19-898-326

  Check Availability
Add to Cart

    Specifications

    Active Ingredients Acetaminophen 500mg
    For Use With (Application) Relieves headaches, muscular aches, pains and fever
    Format Tablet
    Inactive Ingredients Corn Starch, Hypromellose, Maltodextrin, Microcrystalline Cellulose, PEG, Povidone, Pregelatinized Starch, Sodium Starch Glycolate, Stearic Acid, Titanium Dioxide
    Quantity 100 Pack
    Recommended Age 12+ Year(s)
    Storage Requirements 15 to 30°C
    Product Type Medi-First™ Extra Strength Non-Aspirin Tablet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit