Learn More
Fisherbrand™ Medium Bench Height Class 100 Clean Room Chair with Chrome Foot Ring and Adjustable Arms Products for Science Education
Fisherbrand Medium Bench Height Class 100 Clean Room Chair with Chrome Foot Ring and Adjustable Arms features antimicrobial/antibacterial vinyl. Fully adjustable for standard 36 in counters.
$392.00 - $420.00
Specifications
Adjustable (Height) 22 to 29 in
Arm Style ADJUSTABLE
Base Style CHROME
Capacity 300 lbs
Footrest YES
View More Specs
Products 20
Catalog Number Color With Casters With Armrest With Tilt Price Quantity & Availability  
Catalog Number Color With Casters With Armrest With Tilt Price Quantity & Availability  
S40032 Products for Science Education View Documents Blue YES YES NO Each for $392.00
Add to cart
S40045 Products for Science Education View Documents Blue NO YES NO Each for $392.00
Add to cart
S40052 Products for Science Education View Documents Blue YES YES YES Each for $420.00
Add to cart
S40057 Products for Science Education View Documents Blue NO YES YES Each for $420.00
Add to cart
S40062 Products for Science Education View Documents BLACK YES YES NO Each for $392.00
Add to cart
S40065 Products for Science Education View Documents BLACK NO YES NO Each for $392.00
Add to cart
S40068 Products for Science Education View Documents BLACK NO YES YES Each for $420.00
Add to cart
S40087 Products for Science Education View Documents BURGUNDY YES YES NO Each for $392.00
Add to cart
S40089 Products for Science Education View Documents BURGUNDY NO YES NO Each for $392.00
Add to cart
S40092 Products for Science Education View Documents BURGUNDY NO YES YES Each for $420.00
Add to cart
S40097 Products for Science Education View Documents Gray YES YES NO Each for $392.00
Add to cart
S40098 Products for Science Education View Documents Gray NO YES NO Each for $392.00
Add to cart
S40108 Products for Science Education View Documents Gray YES YES YES Each for $420.00
Add to cart
S40112 Products for Science Education View Documents Gray NO YES YES Each for $420.00
Add to cart
S40148 Products for Science Education View Documents Navy NO YES NO Each for $392.00
Add to cart
S40158 Products for Science Education View Documents Navy NO YES YES Each for $420.00
Add to cart
S40067 Products for Science Education View Documents BLACK YES YES YES Each for $420.00
Add to cart
S40090 Products for Science Education View Documents BURGUNDY YES YES YES Each for $420.00
Add to cart
S40147 Products for Science Education View Documents Navy YES YES NO Each for $392.00
Add to cart
S40150 Products for Science Education View Documents Navy YES YES YES Each for $420.00
Add to cart
Description
Description
  • Features ergonomic adjustments
  • Larger Back Pad for extra comfort
  • Optional 8-degree forward, 3-degreee backward locking seat tilt
  • Optional 6-way adjustable arms for proper ergonomic arm placement
  • For 36 in high work surface
  • 20-Inch diameter chrome foot ring
  • Suitable for Class 100 (ISO Class 5) Clean Rooms
  • Antimicrobial/antibacterial vinyl for easy cleaning
  • Stationary glides or self-braking, dual-wheel casters suitable for hard floors
Certifications
ANSI/BIFMA
Warranty and Services
15-year on parts, 5-year on upholstery, Lifetime on the pneumatic lift
Specifications
Specifications
22 to 29 in
CHROME
YES
MEDIUM BENCH
ADJUSTABLE
300 lbs
MEDIUM
YES