Learn More
Fisherbrand™ Medium Bench Height Class 100 Clean Room/ESD Chair with Medium Back Products for Science Education
Fisherbrand Medium Bench Height Class 100 Clean Room/ESD Chair with Medium Back features electro-static dissipating components for highly sensitive applications. Fully adjustable for standard 36 in counters.
$440.00 - $468.00
Specifications
Adjustable (Height) 22 to 29 in
Arm Style Not adjustable
Base Style CHROME
Capacity 300 lbs
Lift Type PNEUMATIC
View More Specs
Products 8
Catalog Number Color With Casters With Tilt Price Quantity & Availability  
Catalog Number Color With Casters With Tilt Price Quantity & Availability  
S39957 Products for Science Education View Documents Blue YES NO Each for $440.00
Add to cart
S39962 Products for Science Education View Documents Blue NO NO Each for $440.00
Add to cart
S39963 Products for Science Education View Documents Black YES NO Each for $440.00
Add to cart
S39968 Products for Science Education View Documents Black NO NO Each for $440.00
Add to cart
S39969 Products for Science Education View Documents Black YES YES Each for $468.00
Add to cart
S39970 Products for Science Education View Documents Black NO YES Each for $468.00
Add to cart
S39971 Products for Science Education View Documents Blue YES YES Each for $468.00
Add to cart
S39972 Products for Science Education View Documents Blue NO YES Each for $468.00
Add to cart
Description
Description
  • Features ergonomic adjustments
  • For 36 in high work surface
  • 20-Inch diameter chrome foot ring
  • Larger Back Pad for extra comfort
  • Optional 8-degree forward, 3-degreee backward locking seat tilt
  • Suitable for Class 100 (ISO Class 5) Clean Rooms
  • Electro-Static Dissipating Vinyl Upholstered Seat and Back
  • Electro-Static Dissipating, Non-Marring Casters or Stationary Glides for proper grounding
Certifications
ANSI/BIFMA, ANSI/ESD-STM-12.1-2006
Warranty and Services
15-year on parts, 5-year on upholstery, Lifetime on the pneumatic lift
Specifications
Specifications
22 to 29 in
CHROME
PNEUMATIC
MEDIUM
YES
Not adjustable
300 lbs
Vinyl
MEDIUM BENCH
YES