Learn More
Brady™ Medium Self-Laminating Blank Tags
Prevent your text from smearing or smudging in some of the harshest environments. Brady™ Medium Self-Laminating Blank Tags are durable one-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 102006
 View more versions of this product
Catalog No. 19-112-948
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are water-resistant
Specifications
Specifications
Green
(25) nylon fasteners
Rectangle
Polyester
3 in. x 3 in.
25/Pack
Water-resistant