Medline SensiCare™ Non-latex Exam Gloves
Disposable, premium polymer vinyl gloves offer a cost-effective, latex-free alternative well suited to the majority of clinical applications
Manufacturer: Medline Industries 484402
 View more versions of this product
Catalog No. 18-999-434B
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Exterior Finish Smooth
    Glove Cuff Style Beaded
    Length (English) 9.5 in.
    Length (Metric) 24.1cm
    Material Vinyl
    Powdered Yes
    Size Medium
    Latex Free Yes
    Sterile Yes
    Thickness 5.1mil
    Color Beige
     Premium vinyl features ultra-highmolecular-weight resins and special softening and strengthening agent
  • Description & Specifications
Related Products
Product Content Feedback