Learn More
Fisherbrand™ Thermal Pouches with Zipper Closure
Fisherbrand Thermal Pouches with Zipper Closure keep temperature sensitive pharmaceutical products like insulin, vaccines and specimens cold for extended periods of time without ice
 View more versions of this product
Catalog No. 07-100-109
Add to cart
Description
Description
Features:
  • Outer reflective surface
  • Inner food grade PR film
  • Micro cell foam insulation
  • Metallized outer film reflects radiant heat
  • Feature a double track zipper for ease of use
  • Use with gel pack, ice or dry ice for longer cooling time
  • Made in USA
Boxes not included
Order Info
Case quantity only 200 per case
Warranty and Services
Warranty against defects in material or workmanship.
Specifications
Specifications
Clear Zipper
7in X 10in
178mm
200
Thermal Mailer
Metalized PET Silver Exterior
12mm
Outer Layer: Metalized PET laminated to PE, Middle Layer: Open Cell PU Foam, Inner Layer: White LDPE (FDA approved for incidental food contact).
245mm