Learn More
Cell Biolabs IncSupplier Diversity Partner MEF FEEDER CELLS PUROMYC  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner CBA312
MEF Feeder Cells are mouse embryonic fibroblasts that allow stem cell culture without LIF. Cells must be mitotically inactivated prior to addition of stem cells. Puromycin resistant.
Catalog No. 50-672-700