Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEF2A Mouse anti-Human, Biotin, Clone: 2F9H2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237298B

Catalog No. NBP237298B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

MEF2A
2F9H2
Western Blot, ELISA
Purified
MEF2A
Purified recombinant fragment of human MEF2A (AA: 391-497) expressed in E. Coli.
Protein A or G purified
Phospho Specific
Primary
4205
ELISA, Western Blot
Biotin
PBS with 0.05% Sodium Azide
ADCAD1, MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancerfactor 2A), MEF2, myocyte enhancer factor 2A, myocyte-specific enhancer factor 2A, RSRFC4, RSRFC9, Serum response factor-like protein 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit