Learn More
MEF2A Mouse anti-Human, DyLight 550, Clone: 2F9H2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237298R
Catalog No. NBP237298R
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MEF2A
2F9H2
Western Blot, ELISA
Purified
MEF2A
Purified recombinant fragment of human MEF2A (AA: 391-497) expressed in E. Coli.
Protein A or G purified
Phospho Specific
Primary
4205
ELISA, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
ADCAD1, MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancerfactor 2A), MEF2, myocyte enhancer factor 2A, myocyte-specific enhancer factor 2A, RSRFC4, RSRFC9, Serum response factor-like protein 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human