Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEF2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA586177
Description
The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.Specifications
MEF2A | |
Polyclonal | |
Unconjugated | |
MEF2A | |
A430079H05Rik, ADCAD1, C5DELq14.3, DEL5q14.3, MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancer factor 2A), MADS box transcription enhancer factor 2, polypeptide C, mef2, Mef2a, MEF2C, myocyte enhancer factor 2a, myocyte enhancer factor 2C, Myocyte-specific enhancer factor 2A, myocyte-specific enhancer factor 2C, RSRFC4, RSRFC9, Serum response factor-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17258, 309957, 4205 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.2 | |
Q02078, Q2MJT0, Q60929 | |
MEF2A | |
Synthetic peptide, corresponding to amino acids 280-330 of Human MEF-2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction