Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEF2A Rabbit anti-Human, Clone: JA33-04, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266943
Description
MEF2A Monoclonal antibody specifically detects MEF2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MEF2A | |
JA33-04 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
ADCAD1, MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancerfactor 2A), MEF2, myocyte enhancer factor 2A, myocyte-specific enhancer factor 2A, RSRFC4, RSRFC9, Serum response factor-like protein 1 | |
Rabbit | |
IgG | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse, Rat |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Purified | |
MEF2A | |
C terminal Human MEF2A | |
Protein A purified | |
Phospho Specific | |
Primary | |
4205 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title