Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEF2B Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $499.50
Specifications
Antigen | MEF2B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MEF2B Polyclonal specifically detects MEF2B in Mouse samples. It is validated for Western Blot.Specifications
MEF2B | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
4207 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
LOC729991-MEF2B readthrough transcript, MEF2B, myocyte enhancer factor 2B | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEF2B (NP_005910.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title