Learn More
MEF2C Mouse anti-Human, Mouse, DyLight 405, Clone: 6H2G2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237427V
Catalog No. NBP237427V
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MEF2C
6H2G2
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
MEF2C
Purified recombinant fragment of human MEF2C (AA: 1-125) expressed in E. Coli.
Protein A or G purified
Neuroscience
Primary
4208
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
DEL5q14.3, myocyte enhancer factor 2C, myocyte-specific enhancer factor 2C, polypeptide C
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse