Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEF2C Mouse anti-Human, Mouse, PE, Clone: 6H2G2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237427PE

Catalog No. NBP237427PE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

MEF2C
6H2G2
Flow Cytometry
Purified
MEF2C
Purified recombinant fragment of human MEF2C (AA: 1-125) expressed in E. Coli.
Protein A or G purified
Neuroscience
Primary
4208
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
DEL5q14.3, myocyte enhancer factor 2C, myocyte-specific enhancer factor 2C, polypeptide C
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit