Learn More
MEF2D 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00004209T01
Catalog No. 89-009-242
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
MEF2D 293T Cell Transient Overexpression Lysate(Denatured)
57.42
pCMV-MEF2D full-length
100 μL
Human
293T
NM_005920.2
6.8
Transient overexpression cell Lysate was tested with Anti-MEF2D antibody (H00004209-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.