Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW™ MegaMan™ Biodegradable Nitrile Gloves with DriTek™ and EcoTek™

Multi-layered nitrile gloves last three times longer than typical single-use nitrile gloves. A fully textured surface enhances grip capabilities, while DriTek™ absorbent lining helps to keep hands cool and dry.

$302.00

Specifications

Applications Industrial, Laboratory
AQL 1.5
Certifications/Compliance FDA 21 CFR 177.2600
Glove Cuff Style Beaded
Color Black
View More Specs

Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-441-062
View Documents
SW™
N260881
X-Small
Case of 500 for $302.00
Add to cart
 
17-441-063
View Documents
SW™
N260882
Small
Case of 500 for $302.00
Add to cart
 
17-441-064
View Documents
SW™
N260883
Medium
Case of 500 for $302.00
Add to cart
 
17-441-065
View Documents
SW™
N260884
Large
Case of 500 for $302.00
Add to cart
 
17-441-066
View Documents
SW™
N260885
X-Large
Case of 500 for $302.00
Add to cart
 
17-441-067
View Documents
SW™
N260886
2X-Large
Case of 500 for $302.00
Add to cart
 
17-441-068
View Documents
SW™
N260887
3X-Large
Case of 500 for $302.00
Add to cart
 
Description

Description

  • EcoTek™ Biodegradability Technology accelerates landfill breakdown without any performance loss
  • DriTek™ Comfort Technology with absorbent lining that wicks away moisture for added extended us
  • ANSI/EN Level 3 abrasion resistance for durability and extended wear
  • Fully textured surface enhances grip capabilities
  • Optimal thickness for durability and tactile sensitivity
  • Gloves last up to 3x longer than standard nitrile gloves
  • 1.5 AQL exceeds laboratory standards
  • SHA Dermatologically Approved
Specifications

Specifications

Industrial, Laboratory
FDA 21 CFR 177.2600
Black
Fully Textured
10.1 mil
8.5 mil
1.5
Beaded
Yes
Yes
Nitrile
MM-11BK
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit