Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Megestrol acetate, ≥99% (HPLC), MilliporeSigma™ Supelco™

Megestrol acetate is a commonly used synthetically prepared glucosteroid, which possesses anti-inflammatory property.

Manufacturer:  MilliporeSigma Supelco M05131G

 View more versions of this product

Catalog No. 11-101-2216


Add to cart

Description

Description

Specifications

Specifications

595-33-5
MFCD00056470
384.51
≥99% (HPLC)
Poly Bottle
Analytical Standard
C24H32O4
17 alpha-Acetoxy-6-methyl-4,6-pregnadiene-3,20-dione; 17 alpha-Hydroxy-6-methyl-4,6-pregnadiene-3,20-dione 17-acetate; 17 alpha-Hydroxy-6-methyl-6-dehydroprogesterone 17-acetate; Megestrol 17-acetate
Neat
Analytical Standard (for drug analysis)
1 g
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit