Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Megestrol acetate, ≥99% (HPLC), MilliporeSigma™ Supelco™

Megestrol acetate is a commonly used synthetically prepared glucosteroid, which possesses anti-inflammatory property.

$130.70 - $418.00

Specifications

Percent Purity ≥99% (HPLC)
Grade Analytical Standard (for drug analysis)
Linear Formula C24H32O4
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-2216
View Documents
MilliporeSigma Supelco
M05131G
1 g
Each for $130.70
Add to cart
 
11-101-2217
View Documents
MilliporeSigma Supelco
M05135G
5 g
Each for $418.00
Add to cart
 
Description

Description

Specifications

Specifications

≥99% (HPLC)
Analytical Standard (for drug analysis)
C24H32O4
Neat
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit