Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGF11 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191360

 View more versions of this product

Catalog No. NBP191360


Add to cart

Description

Description

MEGF11 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

MEGF11
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
MEGF11
Synthetic peptide directed towards the middle region of human MEGF11. Peptide sequence LMMEELNPYTKISPALGAERHSVGAVTGIMLLLFLIVVLLGLFAWHRRRQ.
Immunogen affinity purified
RUO
Primary
84465
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_115821
DKFZp434L121, KIAA1781, multiple EGF-like domains protein 11, multiple EGF-like-domains 11, multiple epidermal growth factor-like domains protein 11
Rabbit
IgG
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Chicken: 92%; Equine: 92%; Zebrafish: 92%.
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit