Learn More
Cole-Parmer Megger 210415 Megohmmeter Model , 15,000 V DC, 300,000 M Ohms  DFS Item
Manufacturer:  Cole-Parmer 2001839
This megohmmeter provides 2% kV and M Ohm accuracy and guard terminal to eliminate surface leakage current from the measurement. It has a zero-start, high-voltage interlock and shielded, high-voltage output cable permanently connected to the instrument. It also has circuit-breaker protection, redundant grounding connections and easy-to-follow, color-coded meter scales, along with analog metering for monitoring of variable measurements and rugged field construction.
Catalog No. 50-631-571