Learn More
Supply SolutionsSupplier Diversity Partner Hach - 500Ml Mehlich 2 Soil Extractant Solution - Platform: Solution; Hazardous: Yes DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 2266349
Hach - 500ml Mehlich 2 Soil Extractant Solution - Platform: Solution; Hazardous: Yes; Parameter: Phosphorus, Acid Hydrolyzable, Phosphorus, Reactive (Ortho), Phosphorus, Total; Extractant for phosphate and potassium determination in soils.; Platform : Solution
Catalog No. 50-476-399