Learn More
Bachem Fmoc-N-Me-Ile-OH, 5g, 138775-22-1, MFCD00153389 DFS Item
Manufacturer:  Bachem 4042030.0005
MeIle derivative used in the Fmoc-SPPS of cyclosporin and compstatin analogs.
Catalog No. 50-194-7217