Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meis homeobox 3 Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: OTI3E12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272642G
Description
Meis homeobox 3 Monoclonal specifically detects Meis homeobox 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Meis homeobox 3 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MEIS3 | |
Recombinant protein fragment corresponding to amino acids 1-261 of human Meis homeobox 3 (NP_064545) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI3E12 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
DKFZp547H236, Meis homeobox 3, Meis1, myeloid ecotropic viral integration site 1 homolog 3, Meis1, myeloid ecotropic viral integration site 1 homolog 3 (mouse), Meis1-related protein 2, MRG2homeobox protein Meis3 | |
Mouse | |
Affinity Purified | |
Neuroscience | |
56917.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title