Learn More
Aviva Systems Biology Corp MEIS3 Antibody - middle region (ARP47650_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MEIS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MEIS3. Alternate names: DKFZp547H236, MRG2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59622