Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEIS3 Mouse anti-Human, Clone: OTI3E12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526416

Catalog No. PIMA526416


Add to cart

Description

Description

Transcriptional regulator which directly modulates PDPK1 expression, thus promoting survival of pancreatic beta-cells. Also regulates expression of NDFIP1, BNIP3, and CCNG1.
Specifications

Specifications

MEIS3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q99687
MEIS3; MRG2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56917
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3E12
Unconjugated
MEIS3
Liquid
MEIS3
Human recombit protein fragment corresponding to amino acids 1-261 of MEIS3 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit