Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ MeisterHand™ Lister Bandage Scissors

Designed for cutting tight bandages

Supplier:  Integra™ Miltex™ MH5514

Catalog No. 12-460-667


Add to Cart

Description

Description

  • Stainless steel
  • Angled blades, extra fine
  • Length: 5.75 in. (14.6cm)
  • Non-glare matte finish

Specifications

Specifications

Lister Bandage Scissors
Standard
13.97 cm
Bandage and cast
Straight
5.5 in.
Stainless Steel
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit