Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK2 Mouse anti-Human, Mouse, Rat, Canine, Monkey, DyLight™ 405, Clone: OTI1A2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272647V
Description
MEK2 Monoclonal specifically detects MEK2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MEK2 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAP2K2 | |
Full length recombinant protein of human MEK2 (NP_109587) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1A2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ERK activator kinase 2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2FLJ26075, p45, PRKMK2MAPKK2 | |
Mouse | |
Affinity Purified | |
Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction | |
5605.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title