Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen MEK2
Applications Immunohistochemistry, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092397
View Documents
Novus Biologicals
NBP302952100UL
100 μg
Each for $320.71
Add to cart
 
NB092398
View Documents
Novus Biologicals
NBP30295220UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

MEK2 Polyclonal antibody specifically detects MEK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, KnockDown
Specifications

Specifications

MEK2
Unconjugated
Rabbit
Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction
PBS with 50% glycerol, pH7.3.
5605
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
ERK activator kinase 2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2FLJ26075, p45, PRKMK2MAPKK2
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MAP2K2 (NP_109587.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit