Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK6 Mouse anti-Human, Clone: 09, Invitrogen™
Mouse Monoclonal Antibody
Specifications
Antigen | MEK6 |
---|---|
Clone | 09 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA542988
![]() |
Thermo Scientific
MA542988 |
100 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
PIMA543511
![]() |
Thermo Scientific
MA543511 |
50 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Specificity: Human MKK6/MEK6/MAP2K6
MEK6 Monoclonal antibody specifically detects MEK6 in Human samples. It is validated for ELISASpecifications
MEK6 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 6; MAP2K6; MAPK/ERK kinase 3; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MAPKK3; MAPKK6; MEK 3; MEK 6; MEK3; MEK6; mitogen activated protein kinase kinase 6; mitogen-activated protein kinase kinase 6; MKK3; MKK6; PRKMK3; Prkmk6; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 2; SAPK kinase 3; SAPKK2; SAPKK-2; SAPKK3; SAPKK-3; SKK3; Stress-activated protein kinase kinase 3 | |
MAP2K6 | |
IgG1 | |
Protein A | |
Antibody |
09 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P52564 | |
5608 | |
Recombinant Human MKK6/MEK6/MAP2K6 protein. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
MAP2K6 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title