Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melan-A/MART-1, Mouse anti-Human,Mouse,Rat,Canine, Biotin, Clone: rMLANA/788, Novus Biologicals™
Mouse Monoclonal Antibody
$627.50
Specifications
Antigen | Melan-A/MART-1 |
---|---|
Clone | rMLANA/788 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB008236
|
Novus Biologicals
NBP254444B0.1ML |
0.1 mL |
Each of 1 for $627.50
|
|
|||||
Description
Melan-A/MART-1 Monoclonal specifically detects Melan-A/MART-1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Melan-A/MART-1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Biotin | |
Mouse | |
Cell Biology, Cellular Markers, Immunology | |
PBS | |
2315 | |
Recombinant full-length human MLANA protein | |
Primary | |
Store at 4°C in the dark. |
rMLANA/788 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Canine | |
Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A | |
MLANA | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title