Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melan-A/MART-1 Rabbit anti-Human, Mouse, Rat, PE-Cyanine7, Clone: MLANA/1761R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254568PECY7

Catalog No. N254568PEC7


Add to cart

Description

Description

Melan-A/MART-1 Monoclonal antibody specifically detects Melan-A/MART-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

Melan-A/MART-1
MLANA/1761R
Flow Cytometry
Conjugated, Purified
MLANA
Recombinant human full-length MLANA protein
Protein A purified
Cytoskeleton Markers, Immunology
Primary
2315.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A
Rabbit
IgG Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit