Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanocortin-1 R/MC1R Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | Melanocortin-1 R/MC1R |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Melanocortin-1 R/MC1R Polyclonal specifically detects Melanocortin-1 R/MC1R in Mouse, Rat samples. It is validated for Western Blot.Specifications
Melanocortin-1 R/MC1R | |
Polyclonal | |
Purified | |
RUO | |
Mouse, Rat | |
CMM5, MC1-R, melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor), Melanocortin receptor 1, melanocyte-stimulating hormone receptor, melanotropin receptor, MSHR, MSH-RMGC14337, SHEP2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human Melanocortin-1 R/MC1R (NP_002377.4). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR, Neuroscience | |
PBS with 50% glycerol, pH7.3. | |
4157 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title