Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanoma Marker (MART-1 + Tyrosinase + gp100) Antibody (M2-7C10 + M2-9E3 + T311 + HMB45), Novus Biologicals™
Mouse Monoclonal Antibody
$265.00 - $506.50
Specifications
Antigen | Melanoma Marker (MART-1 + Tyrosinase + gp100) |
---|---|
Clone | M2-7C10 + M2-9E3 + T311 + HMB45 |
Dilution | Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP234337F
|
Novus Biologicals
NBP2343370.02MG |
0.02 mg |
Each for $265.00
|
|
|||||
NBP234337A
|
Novus Biologicals
NBP2343370.1MG |
0.1 mg |
Each for $506.50
|
|
|||||
NBP234337B
|
Novus Biologicals
NBP2343370.2MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Melanoma Marker (MART-1 + Tyrosinase + gp100) Monoclonal specifically detects Melanoma Marker (MART-1 + Tyrosinase + gp100) in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Melanoma Marker (MART-1 + Tyrosinase + gp100) | |
Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
2315 | |
IgG2b, IgG2a, IgG1 | |
Protein A purified | |
This antibody cocktail recognizes three melanoma-specific proteins, which include MART-1, Tyrosinase and gp100. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients. Function of gp100 is not known but it is reported to be a useful marker for melanocytes and melanomas. This cocktail of three markers is designed for extremely sensitive labeling of formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation. |
M2-7C10 + M2-9E3 + T311 + HMB45 | |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human | |
Q16655 | |
Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45) | |
Primary | |
Store at 4C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title