Learn More
Melanoma Mouse anti-Human, Clone: KBA.62, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB14429
Catalog No. 89-034-512
Add to cart
Description
Description
Mouse monoclonal antibody raised against human melanoma.
  • High specificity
  • High reactivity
Specifications
Specifications
Melanoma
KBA.62
Mouse monoclonal antibody raised against human melanoma.
In 10 mM PBS (0.05% BSA and 0.05% azide).
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Human KAL cells derived from lymph node metastasis of malignant melanoma.
Protein A/G purification
RUO
Primary
Reacts positively against melanocytic tumors but not other tumors.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)