Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melting Point Standard (69-71°C), MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 774025G
Description
Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal CareSpecifications
5 g | |
Analytical Standard | |
MFCD00002752 | |
284.48 | |
1 mmHg (173.7°C) | |
361°C | |
Neat |
57-11-4 | |
CH3(CH2)16 COOH | |
Stearic acid; 1-Heptadecanecarboxylic acid; C18:0; Cetylacetic acid; NSC 25956; NSC 261168; Octadecanoic acid; Stearophanic acid | |
Analytical Standard | |
67°C to 72°C; 69°C to 71°C (±0.3 C) | |
113°C - Closed Cup |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
RTECSNumber : WI2800000
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction