Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEM chloride, 94%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
3970-21-6,542-88-1
C4H9ClO2
124.57
MFCD00000888
BIAAQBNMRITRDV-UHFFFAOYSA-N
2-methoxyethoxymethyl chloride,1-chloromethoxy-2-methoxyethane,ethane, 1-chloromethoxy-2-methoxy,mem chloride,methoxyethoxymethyl chloride,mem-chloride,1-chloro-2,5-dioxahexane,2-methoxyethoxy methyl chloride,beta-methoxyethoxymethyl chloride,.beta.-methoxyethoxymethyl chloride
77590
1-(chloromethoxy)-2-methoxyethane
COCCOCCl
3970-21-6,542-88-1
C4H9ClO2
124.57
MFCD00000888
BIAAQBNMRITRDV-UHFFFAOYSA-N
2-methoxyethoxymethyl chloride,1-chloromethoxy-2-methoxyethane,ethane, 1-chloromethoxy-2-methoxy,mem chloride,methoxyethoxymethyl chloride,mem-chloride,1-chloro-2,5-dioxahexane,2-methoxyethoxy methyl chloride,beta-methoxyethoxymethyl chloride,.beta.-methoxyethoxymethyl chloride
77590
1-(chloromethoxy)-2-methoxyethane
COCCOCCl
Density 1.0900g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 50.0°C to 52.0°C (13.0mmHg)
Refractive Index 1.4260 to 1.4280
Quantity 25g
Color Colorless to Yellow
Specific Gravity 1.09
Flash Point 54°C
Assay Percent Range 92% min. (GC)
Linear Formula CH3OCH2CH2OCH2Cl
Fieser 07,227; 09,299
Formula Weight 124.57
Physical Form Liquid
Percent Purity 94%
Chemical Name or Material MEM chloride
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause cancer.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.<

GHS Signal Word: Danger

EINECSNumber : 223-589-8