Learn More
Cytiva Membranes
Manufacturer:  Cytiva 10409870
Catalog No. 09-927-288
Add to cart
Description
Description
Specifications
Specifications
0.6 μm
Membrane
100Each
Sterile
These membranes are used to quantitatively evaluate microorganism counts. The liquid is filtered through a membrane and microorganisms collect on the membrane surface.