Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Membrin Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303314100UL
Description
Membrin Polyclonal specifically detects Membrin in Human samples. It is validated for Western Blot.Specifications
Membrin | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
golgi SNAP receptor complex member 2, GS27Bos1,27 kDa Golgi SNARE protein, membrin | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human Membrin (NP_004278.2). | |
100 μL | |
Membrane Trafficking and Chaperones | |
9570 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction