Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Membrin Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Membrin
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095439
View Documents
Novus Biologicals
NBP303314100UL
100 μg
Each for $320.71
Add to cart
 
NB095440
View Documents
Novus Biologicals
NBP30331420UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Membrin Polyclonal antibody specifically detects Membrin in Human samples. It is validated for Western Blot
Specifications

Specifications

Membrin
Unconjugated
Rabbit
Membrane Trafficking and Chaperones
PBS with 50% glycerol, pH7.3.
9570
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human
golgi SNAP receptor complex member 2, GS27Bos1,27 kDa Golgi SNARE protein, membrin
Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human Membrin (NP_004278.2).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit