Learn More
Molecular DimensionsSupplier Diversity Partner MEMGOLD2 MEMBRANE CRYSTAL  DFS Item
Manufacturer:  Molecular DimensionsSupplier Diversity Partner MD163
Protein Crystallography. MemGold2 screen (96 x 10mL): features 96 of the most recent alpha helical membrane protein crystallization conditions data mined from the PDB. For Prokaryotic and Eukaryotic alpha helical membrane proteins. Wide range of pH's (4 - 10). Small MW PEGs.
Catalog No. NC0624218